5-Methylfuran-2-boronsäurepinakolester 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,4,5,5-Tetramethyl-2-(5-methylfuran-2-yl)-1,3,2-dioxaborolane, 2-Methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)furan, (5-Methylfuran-2-yl)boronic acid Pinacol ester

OR7474-25G
APOSOR7474-25GEA 74.1 EUR
APOSOR7474-25G
5-Methylfuran-2-boronsäurepinakolester 95%
5-Methylfuran-2-boronsäurepinakolester

Remainder pinacol

Formel: C₁₁H₁₇BO₃
Molecular Weight: 208,06 g/mol
Schmelzpunkt: 217 °C
Dichte: 1,023 g/cm³ (25 °C)
Lagertemperatur: Kühlschrank
MDL: MFCD03094689
CAS-Nummer: 338998-93-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR