5-(4-Methoxyphenyl)-2-furoic acid

Lieferant: Apollo Scientific
Achtung

OR7480-1G OR7480-5G
APOSOR7480-1GEA 43.7 EUR
APOSOR7480-1G APOSOR7480-5G
5-(4-Methoxyphenyl)-2-furoic acid
5-(4-Methoxyphenyl)-2-furoic acid
Formel: C₁₂H₁₀O₄
Schmelzpunkt: 182…186 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD03937477
CAS-Nummer: 52938-99-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR