1,2,3-Benzothiadiazole-5-carboxylic acid
Lieferant: Apollo Scientific
|
Achtung
|
OR7489-1G
OR7489-250MG
APOSOR7489-1GEA
132
EUR
APOSOR7489-1G
APOSOR7489-250MG
1,2,3-Benzothiadiazole-5-carboxylic acid
1,2,3-Benzothiadiazole-5-carboxylic acid
Formel:
C₇H₄N₂O₂S Molecular Weight: 180.19 g/mol Schmelzpunkt: 221…225 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01570552 CAS-Nummer: 192948-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...