4'-Methylbiphenyl-4-carboxaldehyde 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4'-Methyl-[1,1'-biphenyl]-4-carbaldehyde

OR7513-1G OR7513-5G
APOSOR7513-1GEA 139 EUR
APOSOR7513-1G APOSOR7513-5G
4'-Methylbiphenyl-4-carboxaldehyde 97%
4'-Methylbiphenyl-4-carboxaldehyde
Formel: C₁₄H₁₂O
Molecular Weight: 196.249 g/mol
Schmelzpunkt: 104…108 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02258938
CAS-Nummer: 36393-42-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR