3'-Methylbiphenyl-3-carbonsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3'-Methyl[1,1'-biphenyl]-3-carboxylic acid

OR7517-1G
APOSOR7517-1GEA 26.1 EUR
APOSOR7517-1G
3'-Methylbiphenyl-3-carbonsäure 97%
3'-Methylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00558892
CAS-Nummer: 158619-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR