2,6-Naphthalindicarbonsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Naphthalin-2,6-dicarbonsäure

OR7563-100G OR7563-25G OR7563-5G
APOSOR7563-100GEA 185 EUR
APOSOR7563-100G APOSOR7563-25G APOSOR7563-5G
2,6-Naphthalindicarbonsäure 98%
2,6-Naphthalindicarbonsäure
Formel: C₁₂H₈O₄
Molecular Weight: 216,19 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004105
CAS-Nummer: 1141-38-4
EINECS: 214-527-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR