4-(3-Thienyl)benzenecarbaldehyde

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Thien-3-yl)benzaldehyde

OR7592-100MG OR7592-250MG
APOSOR7592-100MGEA 69.6 EUR
APOSOR7592-100MG APOSOR7592-250MG
4-(3-Thienyl)benzenecarbaldehyde
4-(3-Thienyl)benzenecarbaldehyde
Formel: C₁₁H₈OS
Molecular Weight: 188.25 g/mol
Schmelzpunkt: 101…102 °C
Lagertemperatur: Kühlschrank
MDL: MFCD04039151
CAS-Nummer: 157730-74-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR