1-Chloro-6,6-dimethyl-2-hepten-4-yne 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chloro-6,6-dimethylhept-2-en-4-yne

OR7607-25G OR7607-5G OR7607-100G
APOSOR7607-25GEA 29 EUR
APOSOR7607-25G APOSOR7607-5G APOSOR7607-100G
1-Chloro-6,6-dimethyl-2-hepten-4-yne 95%
1-Chloro-6,6-dimethyl-2-hepten-4-yne
Formel: C₉H₁₃Cl
Molecular Weight: 156,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD04039160
CAS-Nummer: 126764-17-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR