4-(Aminomethyl)phenol
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Hydroxybenzylamine
OR7611-5G
OR7611-25G
OR7611-100G
APOSOR7611-5GEA
21.9
EUR
APOSOR7611-5G
APOSOR7611-25G
APOSOR7611-100G
4-(Aminomethyl)phenol
4-(Aminomethyl)phenol
Formel:
C₇H₉NO Molecular Weight: 123.16 g/mol Schmelzpunkt: 116…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD19690971 CAS-Nummer: 696-60-6 UN: 3259 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...