1,2-Dimethyl-1H-indole-3-carboxaldehyde 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2-Dimethyl-1H-indole-3-carbaldehyde
OR7637-1G
OR7637-250MG
APOSOR7637-1GEA
29
EUR
APOSOR7637-1G
APOSOR7637-250MG
1,2-Dimethyl-1H-indole-3-carboxaldehyde 95+%
1,2-Dimethyl-1H-indole-3-carboxaldehyde
Formel:
C₁₁H₁₁NO Molecular Weight: 173,22 g/mol Schmelzpunkt: 127…129 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00745294 CAS-Nummer: 38292-40-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...