4-tert-Butylphenylacetic acid 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(4-(tert-Butyl)phenyl)acetic acid

OR7658-1G OR7658-25G OR7658-5G
APOSOR7658-1GEA 23.4 EUR
APOSOR7658-1G APOSOR7658-25G APOSOR7658-5G
4-tert-Butylphenylacetic acid 97+%
4-tert-Butylphenylacetic acid
Formel: C₁₂H₁₆O₂
Molecular Weight: 192,26 g/mol
Schmelzpunkt: 77…78 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082593
CAS-Nummer: 32857-63-9
EINECS: 251-264-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR