6-Chlor-3-methyl-2,4(1H,3H)-pyrimidindion
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Chloro-3-methylpyrimidine-2,4(1H,3H)-dione
OR7663-1G
OR7663-25G
OR7663-5G
APOSOR7663-1GEA
46.4
EUR
APOSOR7663-1G
APOSOR7663-25G
APOSOR7663-5G
6-Chlor-3-methyl-2,4(1H,3H)-pyrimidindion
6-Chloro-3-methyluracil
Formel:
C₅H₅ClN₂O₂ Molecular Weight: 160.56 g/mol Schmelzpunkt: 278…280 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01074837 CAS-Nummer: 4318-56-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...