1,3,5-Trichlor-2-iodbenzol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,4,6-Trichloroiodobenzene
OR7691-10G
OR7691-5G
APOSOR7691-10GEA
371
EUR
APOSOR7691-10G
APOSOR7691-5G
1,3,5-Trichlor-2-iodbenzol
1,3,5-Trichlor-2-iodbenzol
Formel:
C₆H₂Cl₃I Molecular Weight: 307,34 g/mol Schmelzpunkt: 52…57 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00052857 CAS-Nummer: 6324-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...