1-[2-Bromo-4-(methylsulphonyl)phenyl]piperazine 95+%

Lieferant: Apollo Scientific

OR7695-1G OR7695-5G
APOSOR7695-1GEA 229 EUR
APOSOR7695-1G APOSOR7695-5G
1-[2-Bromo-4-(methylsulphonyl)phenyl]piperazine 95+%
1-[2-Bromo-4-(methylsulphonyl)phenyl]piperazine
Formel: C₁₁H₁₅BrN₂O₂S
Schmelzpunkt: 139…140 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037932
CAS-Nummer: 849035-69-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR