4-(tert-Butoxycarbonylamino)benzylamine 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Aminobenzylamine, 4-BOC protected, 4-(Boc-amino)benzylamine, tert-Butyl N-[4-(aminomethyl)phenyl]carbamate

OR7699-250MG OR7699-1G OR7699-5G
APOSOR7699-250MGEA 21.9 EUR
APOSOR7699-250MG APOSOR7699-1G APOSOR7699-5G
4-(tert-Butoxycarbonylamino)benzylamine 95+%
4-(tert-Butoxycarbonylamino)benzylamine
Formel: C₁₂H₁₈N₂O₂
Molecular Weight: 222,29 g/mol
Schmelzpunkt: 107…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02183573
CAS-Nummer: 220298-96-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR