N-Acetyl-D-galactosamine

Lieferant: Apollo Scientific

Synonyme: 2-Acetamido-2-deoxy-D-galactose, D-N-Acetylgalactosamine, 2-Acetamino-2-deoxy-alpha-D-galactopyranose, GalNAc, N-Acetylchondrosamine

OR7700T-1G OR7700T-5G OR7700T-10G OR7700T-25G
APOSOR7700T-1GEA 0 EUR
APOSOR7700T-1G APOSOR7700T-5G APOSOR7700T-10G APOSOR7700T-25G
N-Acetyl-D-galactosamine
N-Acetyl-D-galactosamine
Formel: C₈H₁₅NO₆
Molecular Weight: 221,21 g/mol
Schmelzpunkt: 160…161 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065372
CAS-Nummer: 1811-31-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR