tert-Butyl-2-(aminomethyl)phenylcarbamate 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Aminomethyl)aniline, 1-BOC protected

OR7704-1G OR7704-5G
APOSOR7704-1GEA 97.3 EUR
APOSOR7704-1G APOSOR7704-5G
tert-Butyl-2-(aminomethyl)phenylcarbamate 95+%
tert-Butyl-2-(aminomethyl)phenylcarbamate
Formel: C₁₂H₁₈N₂O₂
Molecular Weight: 222.29 g/mol
Dichte: 1.054 g/cm³ (25 °C)
Flammpunkt: 71.7 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037902
CAS-Nummer: 849020-94-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR