4-[(4-Chlorophenyl)sulphonyl]piperidine hydrochloride 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7708-1G
OR7708-5G
APOSOR7708-1GEA
285
EUR
APOSOR7708-1G
APOSOR7708-5G
4-[(4-Chlorophenyl)sulphonyl]piperidine hydrochloride 95+%
4-[(4-Chlorophenyl)sulphonyl]piperidine hydrochloride
Formel:
C₁₁H₁₅Cl₂NO₂S Schmelzpunkt: 278…283 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04115824 CAS-Nummer: 101768-64-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...