5-(4-Methoxyphenyl)isoxazole

Lieferant: Apollo Scientific
Achtung

OR7712-1G OR7712-5G
APOSOR7712-1GEA 109 EUR
APOSOR7712-1G APOSOR7712-5G
5-(4-Methoxyphenyl)isoxazole
5-(4-Methoxyphenyl)isoxazole
Formel: C₁₀H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD02183545
CAS-Nummer: 3672-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR