1-[4-Chloro-2-(methylsulphonyl)phenyl]piperazine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7716-1G
OR7716-5G
APOSOR7716-1GEA
229
EUR
APOSOR7716-1G
APOSOR7716-5G
1-[4-Chloro-2-(methylsulphonyl)phenyl]piperazine 95+%
1-[4-Chloro-2-(methylsulphonyl)phenyl]piperazine
Formel:
C₁₁H₁₅ClN₂O₂S Molecular Weight: 274.77 g/mol Schmelzpunkt: 138…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037931 CAS-Nummer: 849035-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...