2,6-Dichloro-4-(methylsulphonyl)aniline 95+%

Lieferant: Apollo Scientific
Achtung

OR7719-1G OR7719-5G
APOSOR7719-1GEA 139 EUR
APOSOR7719-1G APOSOR7719-5G
2,6-Dichloro-4-(methylsulphonyl)aniline 95+%
2,6-Dichloro-4-(methylsulphonyl)aniline
Formel: C₇H₇Cl₂NO₂S
Molecular Weight: 240.11 g/mol
Schmelzpunkt: 155…156 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00007678
CAS-Nummer: 80866-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR