2,6-Dimethyl-3-(methylsulphonyl)aniline 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7728-1G
OR7728-5G
OR7728-100MG
APOSOR7728-1GEA
214
EUR
APOSOR7728-1G
APOSOR7728-5G
APOSOR7728-100MG
2,6-Dimethyl-3-(methylsulphonyl)aniline 95+%
2,6-Dimethyl-3-(methylsulphonyl)aniline
Formel:
C₉H₁₃NO₂S Schmelzpunkt: 103…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037921 CAS-Nummer: 10311-40-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...