4'-Iodacetophenon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
p-Iodacetophenon, 1-(4-Iodphenyl)ethan-1-on, 1-(4-Iodphenyl)ethanon
OR7749-100G
OR7749-25G
OR7749-5G
APOSOR7749-100GEA
65.4
EUR
APOSOR7749-100G
APOSOR7749-25G
APOSOR7749-5G
4'-Iodacetophenon
4'-Iodacetophenon
Formel:
C₈H₇IO Molecular Weight: 246,05 g/mol Schmelzpunkt: 82…84 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00045320 CAS-Nummer: 13329-40-3 EINECS: 236-372-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...