2-Bromphenylhydraziniumchlorid 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2-Bromphenylhydrazin Hydrochlorid, 1-(2-Bromphenyl)hydrazin Hydrochlorid, 2-Bromphenylhydrazinhydrochlorid
OR7758-100G
OR7758-250G
OR7758-25G
APOSOR7758-100GEA
66.7
EUR
APOSOR7758-100G
APOSOR7758-250G
APOSOR7758-25G
2-Bromphenylhydraziniumchlorid 98%
2-Bromphenylhydraziniumchlorid
Formel:
C₆H₈BrClN₂ Molecular Weight: 223,5 g/mol Schmelzpunkt: 182…189 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012926 CAS-Nummer: 50709-33-6 EINECS: 256-728-6 UN: 1759 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...