2-Amino-6-brompyridin 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Brompyridin-2-amin, 6-Brom-2-pyridylamin
OR7768-25G
OR7768-5G
OR7768-1G
APOSOR7768-25GEA
31.9
EUR
APOSOR7768-25G
APOSOR7768-5G
APOSOR7768-1G
2-Amino-6-brompyridin 98%
2-Amino-6-brompyridin
Formel:
C₅H₅BrN₂ Molecular Weight: 173,01 g/mol Schmelzpunkt: 88…91 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00137843 CAS-Nummer: 19798-81-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...