4-Bromphenylboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Dihydroxy(4-bromphenyl)boran, 4-Brombenzolboronsäure, (4-Bromphenyl)metaborsäure
OR7775-25G
OR7775-5G
OR7775-100G
APOSOR7775-25GEA
30.6
EUR
APOSOR7775-25G
APOSOR7775-5G
APOSOR7775-100G
4-Bromphenylboronsäure
4-Bromphenylboronsäure
Formel:
C₆H₆BBrO₂ Molecular Weight: 200,83 g/mol Schmelzpunkt: 282…288 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002104 CAS-Nummer: 5467-74-3 EINECS: 226-779-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...