6-(Methylsulphonyl)-1H-indole 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR7793-1G
OR7793-5G
APOSOR7793-1GEA
1040
EUR
APOSOR7793-1G
APOSOR7793-5G
6-(Methylsulphonyl)-1H-indole 95+%
6-(Methylsulphonyl)-1H-indole
Formel:
C₉H₉NO₂S Molecular Weight: 195.24 g/mol Schmelzpunkt: 147…151 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04037939 CAS-Nummer: 467461-40-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...