4-(Methylsulfonyl)-2-nitroanilin
Lieferant: Apollo Scientific
|
Achtung
|
OR7794-1G
OR7794-10G
OR7794-5G
APOSOR7794-1GEA
62.5
EUR
APOSOR7794-1G
APOSOR7794-10G
APOSOR7794-5G
4-(Methylsulfonyl)-2-nitroanilin
4-(Methylsulfonyl)-2-nitroanilin
Formel:
C₇H₈N₂O₄S Schmelzpunkt: 194…196 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00718367 CAS-Nummer: 21731-56-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...