N-[4-(Methylsulphonyl)-2-nitrophenyl]piperidin-4-amine 95+%
Lieferant: Apollo Scientific
OR7797-1G
OR7797-5G
OR7797-25G
APOSOR7797-1GEA
229
EUR
APOSOR7797-1G
APOSOR7797-5G
APOSOR7797-25G
N-[4-(Methylsulphonyl)-2-nitrophenyl]piperidin-4-amine 95+%
N-[4-(Methylsulphonyl)-2-nitrophenyl]piperidin-4-amine
Formel:
C₁₂H₁₇N₃O₄S Molecular Weight: 299,35 g/mol Schmelzpunkt: 178…179 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04037967 CAS-Nummer: 849035-94-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...