N-Acetyl-β-D-glucosamin 95%
Lieferant: Apollo Scientific
Synonyme:
2-Acetamido-2-deoxy-D-glucopyranose, 2-Acetamido-2-deoxy-D-glucose, D-GlcNAc, N-Acetyl-D-glucosamine
OR7800T-100G
OR7800T-1KG
OR7800T-500G
APOSOR7800T-100GEA
75.4
EUR
APOSOR7800T-100G
APOSOR7800T-1KG
APOSOR7800T-500G
N-Acetyl-β-D-glucosamin 95%
N-Acetyl-β-D-glucosamin
Formel:
C₈H₁₅NO₆ Molecular Weight: 221,21 g/mol Schmelzpunkt: 211 °C (Zersetzung) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00136044 CAS-Nummer: 7512-17-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...