5-Brom-2-methoxypyrimidin 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR7810-5G
OR7810-100G
APOSOR7810-5GEA
0
EUR
APOSOR7810-5G
APOSOR7810-100G
5-Brom-2-methoxypyrimidin 98%
5-Bromo-2-methoxypyrimidine
Formel:
C₅H₅BrN₂O Molecular Weight: 189,01 g/mol Schmelzpunkt: 55…60 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973351 CAS-Nummer: 14001-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...