3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(1,2,3,6-Tetrahydro-4-pyridyl)indole
OR7812-1G
OR7812-250MG
APOSOR7812-1GEA
157
EUR
APOSOR7812-1G
APOSOR7812-250MG
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole 95+%
3-(1,2,3,6-tetrahydropyridin-4-yl)-1H-indole
Formel:
C₁₃H₁₄N₂ Molecular Weight: 198.269 g/mol Schmelzpunkt: 177…182 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03695471 CAS-Nummer: 65347-55-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...