4-[(1-Acetyl-2,3-dihydro-1H-indol-7-yl)amino]-4-oxobutanoic acid 95+%
Lieferant: Apollo Scientific
OR7816-10G
OR7816-1G
OR7816-5G
APOSOR7816-10GEA
1060
EUR
APOSOR7816-10G
APOSOR7816-1G
APOSOR7816-5G
4-[(1-Acetyl-2,3-dihydro-1H-indol-7-yl)amino]-4-oxobutanoic acid 95+%
4-[(1-Acetyl-2,3-dihydro-1H-indol-7-yl)amino]-4-oxobutanoic acid
Formel:
C₁₄H₁₆N₂O₄ Schmelzpunkt: 151…155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00232838 CAS-Nummer: 394654-07-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...