5-Bromo-3-methylindole 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Bromo-3-methyl-1H-indole
OR7820-1G
OR7820-25G
OR7820-5G
APOSOR7820-1GEA
120
EUR
APOSOR7820-1G
APOSOR7820-25G
APOSOR7820-5G
5-Bromo-3-methylindole 98%
5-Bromo-3-methylindole
Formel:
C₉H₈BrN Molecular Weight: 210,08 g/mol Schmelzpunkt: 76…82 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD04038746 CAS-Nummer: 10075-48-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...