1-(5-Bromopyrimidin-2-yl)piperidine-3-carboxylicacid 95+%

Lieferant: Apollo Scientific
Achtung

OR7825-1G OR7825-500MG
APOSOR7825-1GEA 114 EUR
APOSOR7825-1G APOSOR7825-500MG
1-(5-Bromopyrimidin-2-yl)piperidine-3-carboxylicacid 95+%
1-(5-Bromopyrimidin-2-yl)piperidine-3-carboxylicacid
Formel: C₁₀H₁₂BrN₃O₂
Molecular Weight: 286,14 g/mol
Schmelzpunkt: 190…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426071
CAS-Nummer: 799283-93-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR