3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-(Methylamino)propanoic acid, N-BOC protected

OR7830-5G OR7830-1G OR7830-25G
APOSOR7830-5GEA 37.7 EUR
APOSOR7830-5G APOSOR7830-1G APOSOR7830-25G
3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid 98%
3-((tert-Butoxycarbonyl)(methyl)amino)propanoic acid
Formel: C₉H₁₇NO₄
Molecular Weight: 203.2400 g/mol
Schmelzpunkt: 59…62 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03490508
CAS-Nummer: 124072-61-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR