(2E)-3-[4-Chloro-2-(methylsulphonyl)phenyl]-acrylic acid 95+%

Lieferant: Apollo Scientific

OR7835-1G OR7835-5G OR7835-25G
APOSOR7835-1GEA 285 EUR
APOSOR7835-1G APOSOR7835-5G APOSOR7835-25G
(2E)-3-[4-Chloro-2-(methylsulphonyl)phenyl]-acrylic acid 95+%
(2E)-3-[4-Chloro-2-(methylsulphonyl)phenyl]-acrylic acid
Formel: C₁₀H₉ClO₄S
Molecular Weight: 260,7 g/mol
Schmelzpunkt: 251…253 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD04037952
CAS-Nummer: 849035-82-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR