3-(2,3-Dihydro-1-benzofuran-5-yl)acrylic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: (E)-3-(2,3-Dihydrobenzofuran-5-yl)acrylic acid

OR7839-1G
APOSOR7839-1GEA 58 EUR
APOSOR7839-1G
3-(2,3-Dihydro-1-benzofuran-5-yl)acrylic acid 95+%
3-(2,3-Dihydro-1-benzofuran-5-yl)acrylic acid
Formel: C₁₁H₁₀O₃
Molecular Weight: 190,2 g/mol
Schmelzpunkt: 170…173 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03002828
CAS-Nummer: 203505-84-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR