4-[5-(Methylsulphonyl)-2,3-dihydro-1H-indol-1-yl]-4-oxobutanoic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-[2,3-Dihydro-5-(methylsulphonyl)-1H-indol-1-yl]-4-oxobutanoic acid
OR7863-1G
OR7863-25G
OR7863-5G
APOSOR7863-1GEA
209
EUR
APOSOR7863-1G
APOSOR7863-25G
APOSOR7863-5G
4-[5-(Methylsulphonyl)-2,3-dihydro-1H-indol-1-yl]-4-oxobutanoic acid 95+%
4-[5-(Methylsulphonyl)-2,3-dihydro-1H-indol-1-yl]-4-oxobutanoic acid
Formel:
C₁₃H₁₅NO₅S Molecular Weight: 297.33 g/mol Schmelzpunkt: 225…228 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01319206 CAS-Nummer: 396105-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...