1,2-Dihydro-4-oxopyrrolo[3,2,1-ij]quinoline-5-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Oxo-1,2-dihydro-4H-pyrrolo[3,2,1-ij]quinolin-5-carboxylic acid

OR7872-1G OR7872-5G OR7872-250MG
APOSOR7872-1GEA 219 EUR
APOSOR7872-1G APOSOR7872-5G APOSOR7872-250MG
1,2-Dihydro-4-oxopyrrolo[3,2,1-ij]quinoline-5-carboxylic acid 95%
1,2-Dihydro-4-oxopyrrolo[3,2,1-ij]quinoline-5-carboxylic acid
Formel: C₁₂H₉NO₃
Molecular Weight: 215.21 g/mol
Schmelzpunkt: >234 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD01567795
CAS-Nummer: 386715-43-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR