4-Oxo-4-(5-oxo-1,4-diazepan-1-yl)butanoic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Oxo-4-(5-oxohomopiperazin-1-yl)butanoic acid

OR7873-1G OR7873-250MG
APOSOR7873-1GEA 244 EUR
APOSOR7873-1G APOSOR7873-250MG
4-Oxo-4-(5-oxo-1,4-diazepan-1-yl)butanoic acid 95+%
4-Oxo-4-(5-oxo-1,4-diazepan-1-yl)butanoic acid
Formel: C₉H₁₄N₂O₄
Molecular Weight: 214,22 g/mol
Schmelzpunkt: 145…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01319265
CAS-Nummer: 397244-77-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR