3-[3-(Pyrazin-2-yl)-1,2,4-oxadiazol-5-yl]propanoic acid 95+%

Lieferant: Apollo Scientific
Achtung

OR7882-1G OR7882-5G OR7882-250MG
APOSOR7882-1GEA 227 EUR
APOSOR7882-1G APOSOR7882-5G APOSOR7882-250MG
3-[3-(Pyrazin-2-yl)-1,2,4-oxadiazol-5-yl]propanoic acid 95+%
3-[3-(Pyrazin-2-yl)-1,2,4-oxadiazol-5-yl]propanoic acid
Formel: C₉H₈N₄O₃
Molecular Weight: 220.19 g/mol
Schmelzpunkt: >155 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD03426086
CAS-Nummer: 849925-05-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR