4-Thiazolcarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Thiazole-4-carboxylic acid
OR7886-25G
OR7886-5G
APOSOR7886-25GEA
87
EUR
APOSOR7886-25G
APOSOR7886-5G
4-Thiazolcarbonsäure
4-Thiazolecarboxylic acid
Formel:
C₄H₃NO₂S Molecular Weight: 129,1371 g/mol Schmelzpunkt: 196…199 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00623589 CAS-Nummer: 3973-08-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...