4-[3-(2-Thienyl)-1,2,4-oxadiazol-5-yl]butanoic acid 95+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[3-(Thien-2-yl)-1,2,4-oxadiazol-5-yl]butanoic acid

OR7887-1G OR7887-5G OR7887-250MG
APOSOR7887-1GEA 277 EUR
APOSOR7887-1G APOSOR7887-5G APOSOR7887-250MG
4-[3-(2-Thienyl)-1,2,4-oxadiazol-5-yl]butanoic acid 95+%
4-[3-(2-Thienyl)-1,2,4-oxadiazol-5-yl]butanoic acid
Formel: C₁₀H₁₀N₂O₃S
Molecular Weight: 238,27 g/mol
Schmelzpunkt: 87…90 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03426087
CAS-Nummer: 849925-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR