Ethyl-2,6-dichloro-5-nitropyrimidine-4-carboxylate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2,6-dichloro-5-nitropyrimidine-4-carboxylate

OR7888-1G OR7888-250MG OR7888-5G
APOSOR7888-1GEA 55.1 EUR
APOSOR7888-1G APOSOR7888-250MG APOSOR7888-5G
Ethyl-2,6-dichloro-5-nitropyrimidine-4-carboxylate
Ethyl-2,6-dichloro-5-nitropyrimidine-4-carboxylate
Formel: C₇H₅Cl₂N₃O₄
Molecular Weight: 266.04 g/mol
Schmelzpunkt: 36…38 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00831638
CAS-Nummer: 54368-61-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR