1-(4-Benzyloxyphenyl)-2-thiourea

Lieferant: Apollo Scientific
Achtung

OR7897-1G OR7897-5G
APOSOR7897-1GEA 161 EUR
APOSOR7897-1G APOSOR7897-5G
1-(4-Benzyloxyphenyl)-2-thiourea
1-(4-Benzyloxyphenyl)-2-thiourea
Formel: C₁₄H₁₄N₂OS
Molecular Weight: 258.34 g/mol
Schmelzpunkt: 191…197 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00060457
CAS-Nummer: 65069-53-6
UN: 3335
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR