1-(2-Cyanophenyl)-4-piperidinecarboxylic acid

Lieferant: Apollo Scientific
Achtung

OR7898-10G OR7898-1G OR7898-5G
APOSOR7898-10GEA 535 EUR
APOSOR7898-10G APOSOR7898-1G APOSOR7898-5G
1-(2-Cyanophenyl)-4-piperidinecarboxylic acid
1-(2-Cyanophenyl)-4-piperidinecarboxylic acid
Formel: C₁₃H₁₄N₂O₂
Molecular Weight: 230,27 g/mol
Schmelzpunkt: 176…178 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08689748
CAS-Nummer: 937601-79-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR