1-(2-Cyanophenyl)piperidine-3-carboxylic acid
Lieferant: Apollo Scientific
OR7900-1G
OR7900-5G
APOSOR7900-1GEA
118
EUR
APOSOR7900-1G
APOSOR7900-5G
1-(2-Cyanophenyl)piperidine-3-carboxylic acid
1-(2-Cyanophenyl)piperidine-3-carboxylic acid
Formel:
C₁₃H₁₄N₂O₂ Schmelzpunkt: 79…80 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD09027711 CAS-Nummer: 942474-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...