5-Benzylhydantoin

Lieferant: Apollo Scientific
Achtung

OR7901T-1G OR7901T-5G
APOSOR7901T-1GEA 219 EUR
APOSOR7901T-1G APOSOR7901T-5G
5-Benzylhydantoin
5-Benzylhydantoin
Formel: C₁₀H₁₀N₂O₂
Schmelzpunkt: 188…189 °C
Lagertemperatur: Raumtemperatur
CAS-Nummer: 3530-82-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR