Ethyl-5-nitro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
OR7911-100MG
OR7911-500MG
APOSOR7911-100MGEA
82.8
EUR
APOSOR7911-100MG
APOSOR7911-500MG
Ethyl-5-nitro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Ethyl-5-nitro-2,6-dioxo-1,2,3,6-tetrahydropyrimidine-4-carboxylate
Formel:
C₇H₇N₃O₆ Molecular Weight: 229.15 g/mol Schmelzpunkt: 165 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01570795 CAS-Nummer: 52047-16-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...